Learn More
EZ-Run™ Protein Gel Solutions, Fisher BioReagents™
Shop All Fisherbrand ProductsDescription
A ready-to-use premixed solution of acrylamide, bisacrylamide, buffer and SDS that enables ultra-fine resolution of protein bands by denaturing PAGE. This novel formulation provides the resolution of a gradient gel with less preparative work than a standard discontinuous gel system.
- Ready-to-pour EZ-Run Protein Gel Solution for 1-D or 2-D PAGE—just add APS and TEMED
- Wide separation range—from 3 to 200kDa on same minigel
- Continuous gel system—no stacking gel necessary
- Proprietary (patent-pending) gel chemistry
EZ-Run Protein Gel Solution comes with an optimized proprietary running buffer; other traditional SDS-PAGE buffers should not be used with the EZ-Run gel solution.
Applications:
Compatible with post-electrophoresis applications, such as Western blotting, MALDI analysis, protein sequencing. Suitable for all commonly used gel staining methods including Coomassie™ blue, silver staining and fluorescent dyes.

Specifications
Specifications
Chemical Name or Material | Protein Gel Solution |
Composition | 10% protein gel solution |
Concentration | 10% |
Quantity | 500 mL |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.