Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Fisher BioReagents™ EZ-Run™ Protein Gel Staining Solution
Shop All Fisherbrand Products
Click to view available options
Quantity:
1 L
4 L
Description
EZ-Run Protein Gel Staining Solution contains Coomassie™ Brilliant Blue G-250. Due to the property of the dye to form colloidal particles, proteins are preferentially stained without significant background staining of the gel matrix.
- Detection is linear over two orders of magnitude with as little as 5ng protein detected
- Sensitivity: 5ng BSA in 10% SDS-PAGE, 1.0mm thick gel, well width 3mm
- Does not contain toxic methanol or acetic acid
- Provides rapid destaining with simple water washes
- Can be reused twice without significant loss of sensitivity
- Tested for staining sensitivity in minigels using purified proteins and Protein Molecular Weight Standards
Specifications
Specifications
| Color | Dark Blue |
| Product Type | Stain |
| Form | Liquid |
| Formulation | Coomassie™ Brilliant Blue G-250 |
| For Use With (Application) | Highly sensitive staining of proteins separated in Polyacrylamide gels |
| Quantity | 1 L |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction