Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Fisher BioReagents™ exACTGene™ dNTPs
Ultrapure dNTPs ideal for use in PCR, DNA labeling, and DNA sequencing
Specifications
Format | Liquid |
---|---|
Content And Storage | -20°C |
Purity | >99% |
Color | Colorless |
For Use With (Application) | PCR, DNA labeling and DNA sequencing |
Catalog Number | Concentration | pH | Product Type | Quantity | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Concentration | pH | Product Type | Quantity | Price | Quantity & Availability | |||
BP2595200
|
10 mM | 7.3 to 7.7 | dNTP Mix | 200 μL | N/A | N/A | N/A | ||
Description
- >99% triphosphate
- Chemically synthesized to meet a high purity standard
- Ensure consistent results and optimum sensitivity for many rigorous applications
- Tested for purity by HPLC, performance in long-template PCR, and absence of DNase, RNase, and protease activity to ensure dependable performance and lot-to-lot consistency
- Ultrapure dNTPs will enhance results for the following procedures: QPCR, high-fidelity PCR, long template (>8kb) PCR, RT-PCR, fluorescent sequencing, cDNA synthesis, Pyrosequencing™, and in vitro mutagenesis
Specifications
Liquid | |
>99% | |
PCR, DNA labeling and DNA sequencing |
-20°C | |
Colorless |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title