Learn More
Description
- Licensed for use in PCR
- Uses a thermostable DNA polymerase that has been encoded by a modified form of Thermus aquaticus DNA polymerase gene that has been inserted into an E. coli host
- Has a 5«→3« exonuclease activity
- Available with two different reaction buffers
- Buffer A contains an optimized final concentration of 15mM MgCl2 for most PCR amplifications
- Buffer B does not contain MgCl2; MgCl2 is provided in a separate vial as a 25mM solution
- Storage Buffer: 20mM Tris-HCl, pH 7.5 (at room temperature), 100mM KCl, 0.1mM EDTA, 1mM DTT, 50% Glycerol, 0.5% Tween™ 20, 0.5% Nonidet P40™
- Recommended Assay Buffer: 10X Assay Buffer A (100mM Tris-HCl, pH 9.0 [at 25°C/77°F]; 500mM KCl; 15mM MgCl2) or 10X Assay Buffer B (100mM Tris-HCl, pH 9.0 [at 25°C/77°F]; 500mM KCl) and 25mM MgCl2 solution in a separate vial
Unit Definition: One unit of enzyme is defined as the amount that will incorporate 10nmol of dNTPs into acid-insoluble material per 30 minutes in a 10-minute incubation at 74°C (165°F) under the assay conditions above
Specifications
Specifications
| Concentration | 5 U/μL |
| Composition | 10X assay buffer A : Tris-HCl 100mM, pH 9 at 25°C, KCl 500mM and MgCl2 15mM solution in a separate vial |
| Content And Storage | -20°C |
| Description | Taq DNA Polymerase |
| Form | Liquid |
| Format | Poly Tube |
| Quantity | 500 Units |
| For Use With (Application) | Standard PCR |
| Storage Buffer | Tris-HCl 20mM, pH 7.5 (at room temperature), KCl 100mM, EDTA 0.1mM, DTT 1mM, Glycerol 50%, Tween™ 20 0.5%, Nonidet™ P40™ 0.5% |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.