Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisher BioReagents™ Leupeptin Hemisulfate

Catalog No. BP26621 Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
1 mg
5 mg
25 mg
100 mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BP26621 1 mg
BP26625 5 mg
BP266225 25 mg
BP2662100 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. BP26621

(Acetyl-Leu-Leu-Arg-al), Valeramide

A biochemical used as an inhibitor for trypsin, plasmin, papain, and cathepsin B.

TRUSTED_SUSTAINABILITY

Specifications

Color White
For Use With (Application) Inhibitor for Trypsin, Plasmin, Papain, Cathepsin B
Content And Storage -20°C
Quantity 1 mg
Product Type Leupeptin Hemisulfate
Molecular Weight (g/mol) 475.60
Form Chunks, Powder
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.