Learn More
Description
(Acetyl-Leu-Leu-Arg-al), Valeramide
A biochemical used as an inhibitor for trypsin, plasmin, papain, and cathepsin B.
Specifications
Specifications
| Color | White |
| For Use With (Application) | Inhibitor for Trypsin, Plasmin, Papain, Cathepsin B |
| Content And Storage | -20°C |
| Quantity | 25 mg |
| Product Type | Leupeptin Hemisulfate |
| Molecular Weight (g/mol) | 475.60 |
| Form | Chunks, Powder |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.