Learn More
Description
- Application: This enzyme is used to lyse E. coli for preparation of plasmid DNA from transformed cell cultures
- Not on TSCA inventory: for research and development use only; not for manufacturing or commercial purposes
- Enzyme Activity: ≈20,000U/mg dry weight
- Solubility: Readily soluble at 5mg/mL in 0.1M KH2PO4 to give colorless solution
Specifications
Specifications
| Color | White |
| For Use With (Application) | Used to lyse E. coli for preparation of plasmid DNA from transformed cell cultures |
| Content And Storage | <0°C |
| Source | Egg white |
| Quantity | 10 g |
| Product Type | Lysozyme |
| Form | Crystalline Powder |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.