Learn More
Description
Four classic media, in convenient, preweighed pouches.
- Preblended growth-promoting components save time and preparation costs
- Reduce formulation errors
- Each pouch contains enough material for 1 liter of liquid media
- No more weighing of dusty powders
- High quality—assured by stringent testing procedures
- LB is used for the culturing of E. coli in molecular biology procedures
- Terrific Broth is a buffered enriched medium used to obtain high yields of plasmid DNA from transformed E. coli
Order Info
Supplied as 10 pouches per box.
Specifications
Specifications
| Form | Solid |
| Grade | Molecular genetics |
| Media Type | LB Agar, Miller |
| Product Type | Nutrient Agar |
| For Use With (Application) | Culturing of Escherichia coli |
| Packaging Type | Pouch |
| Quantity | 10 Pouches |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.