Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisher BioReagents™ SurePrep™ RNA Cleanup and Concentration Kit

Catalog No. BP280950 Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
50 Preps
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BP280950 50 Preps
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BP280950

missing translation for 'validMainframeMsg'

Includes

Binding solution (2 x 20mL), Wash solution (22mL), RNA elution buffer (6mL), Collection tubes (50), Spin columns (50), Elution tubes (1.7mL) (50), Product insert

Provides a rapid method to purify and concentrate RNA isolated using other methods such as phenol:chloroform extraction or after enzymatic reactions

  • Fast and easy processing using rapid spin-column format
  • Suitable for all sizes of RNA, from large mRNA to microRNA (miRNA)
  • Able to clean up RNA isolated using different methods, including phenol/chloroform extractions
  • Efficient RNA cleanup from various enzymatic reactions such as labeling, DNase treatment and in vitro transcription

Purified RNA is suitable for Quantitative RT-PCR, Northern blotting, RNase protection, primer extension and expression array assays

TRUSTED_SUSTAINABILITY

Specifications

Format Spin Column
For Use With (Application) RT-PCR, northern blotting, RNase protection, primer extension and expression array assays
Includes Binding solution 2 x 20mL, wash solution 22mL, RNA Elution Buffer 6mL, collection tubes, spin columns, elution tubes 1.7mL, product insert
Quantity 50 Preps
Yield 15μg RNA HeLa cells (1 x 106 cells), 8μg DNA HeLa cells (1 x 106 cells), 150μg Protein HeLa cells (1 x 106 cells)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.