Learn More
Fisher Chemical™ Harris Modified Method Hematoxylin Stains
Catalog No. p-121926 Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
4 L
Includes:
Without Acetic Acid
Catalog No. Quantity Includes
SH30-4D 4 L Without Acetic Acid
1 options
Catalog No. SH30-4D
Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.
View Alternate Product
For histological and cytological use as a nuclear stain
  • Mercury-free
  • Suitable for PAP staining
Exposure to low temperatures may cause precipitation of dye. Mix at ambient temperature before use and filter if necessary. Sedimentation does not affect use of this stain.
TRUSTED_SUSTAINABILITY
Specifications
Product Type Hematoxylin
Quantity 4 L
For Use With (Application) PAP Staining
Includes Without Acetic Acid