Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisher Container™ Precision Stat Anti-Static Pink Cleanroom Bags

Catalog No. p-4706488 Shop All Fisher Container Corp. Products
Click to view available options
Dimensions (L x W):
10 x 12 in.

Specifications

Color Pink
For Use With (Application) Sterilization by gamma radiation or ethylene oxide
Thickness (English) 6 mil
Dimensions (L x W) 10 x 12 in.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.