Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisher Healthcare™ Pinnacle Portfolio Cyto-Stain™

Catalog No. 22220105 Shop All Fisherbrand Products
Click to view available options
Type:
Cytostain
Cytostain G
This item is not returnable. View return policy
This item is not returnable. View return policy

Can be used for all gynecological and non-gynecological procedures

Cyto-Stain Features:

  • Uniquely formulated cytoplasmic stain produces results similar to those of conventional Papanicolaou stains
  • Provides a full range of familiar cytoplasmic colors and allows the user control over staining intensities
  • Reduces staining time; cost effective because you need only one counterstain, not two
  • Staining characteristics are comparable to conventional EA-OG stains
  • Cyto-Stain G produces greener cyanophilic hues in intermediate and basal cells

Pinnacle Portfolio

  • Designed as a system
  • Each component has added value and benefits that reduce cost per slide
  • Unique chemical formulae of the promote long-term chemical stability, which extends the usable life of each component
  • Are subjected to some of the highest quality manufacturing standards in the industry
  • Packaging improvements enhance product longevity

Specifications

Type Cytostain G
Format Liquid
For Use With (Application) Can be used for all gynecological and non-gynecological procedures
Quantity 4 x 500mL
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.