Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisher Healthcare™ PROTOCOL™ Histological/Cytological Stains

Catalog No. 23245679 Shop All Fisherbrand Products
Click to view available options
Quantity:
475 mL
500 mL
32 oz.
1 L
1 gal.
4 L
Type:
Alcoholic Eosin Y Stain
Bluing Reagent
Cyto-Quik Combo EA/OG Stain
Eosin Y Intensified
Gill Hematoxylin (1X)
Gill Hematoxylin (2X)
Gill Hematoxylin (3X)
Gomori Trichrome
Harris Hematoxylin
Harris Hematoxylin - Mercury-free
Harris Hematoxylin - Mercury-free, Non-acidified
This item is not returnable. View return policy
This item is not returnable. View return policy

Manufactured with strict quality control to ensure lot-to-lot uniformity

Prepared with stain certified by the Biological Stain Commission

Specifications

Type Gomori Trichrome
Format Liquid
For Use With (Application) Used for Histological/Cytological Staining
Quantity 500 mL
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.