Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisher Healthcare™ PROTOCOL™ Stains for HEMA-TEK™ Automated Slide Stainers

Catalog No. 23036222 Shop All Fisherbrand Products
Click to view available options
Quantity:
5 Case
Each
Color:
Dark Blue
Dark Purple
This item is not returnable. View return policy
This item is not returnable. View return policy

Allow varied staining times to suit individual preferences

Stains for manual and automated staining methods

  • All PROTOCOL Hematology Stains are prepared with materials certified by the Biological Stain Commission to ensure purity
  • Manufactured with strict quality control measures to ensure lot-to-lot uniformity, including extensive analytical and performance testing on human specimens
  • Carefully balanced for optimum staining pH
  • Remain stable for a minimum of 12 months when stored at room temperature
  • Packaged in durable plastic bottles for safety and easy handling
  • Convenient finger tabs and clear reagent identification eliminate improper line connection

Specifications

Quantity Each
Description Stain, Wright's Stain
Storage Requirements Room Temperature
Boiling Point 37.7°C
Format Liquid
For Use With (Equipment) Hema-Tek™ automated slide stainer
Color Dark Purple
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.