Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Fisherbrand™ Sure-Vue™ Signature Rubella Latex Test
Shop All Fisherbrand Products
Click to view available options
Quantity:
100 Tests
500 Tests
Description
Includes
Latex reagent, high positive control, low positive control, negative control, diluent, disposable test cards, and package insert
- CLIA Complexity: Non-Waived
- CPT Codes: 86762/86406
- Sensitivity: 100%
- Specificity: 98.6%
- Rapid latex agglutination test for the qualitative and semi-quantitative detection of both IgG and IgM anti-rubella virus antibodies in serum
- Results in five minutes
- For in vitro diagnostic use
Specifications
Specifications
| CPT Code | 86762, 86406 |
| Clia Complexity | Non-waived |
| Detectable Analytes | Anti-rubella virus IgG, IgM |
| Type | Rubella Latex Test Kit |
| Includes | Latex reagent, high positive control, low positive control, negative control, diluent, disposable test cards and package insert |
| Quantity | 500 Tests |
| Test Time | 5 min. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction