Learn More
Fisher Science Education™ Advanced Digital Stereomicroscopes
SCI_ED
Catalog No. p~4701474
Change view
Click to view available options
Magnification Power:
10X to 30X
Catalog No. Magnification Power
S71012B 10X to 30X
1 options
Catalog No. S71012B Supplier Fisher Science Education™ Supplier No. DC4456H
Only null left
Add to Cart
Add to Cart
Includes
Motic™ Images software, which allows the user to view, capture, label, manipulate and measure images; vinyl dustcover; rubber eyeshields
Extra-rugged stereomicroscopes have a built-in high-resolution digital camera.
College/University Level
  • Versatile stereomicroscope permits user to change from a widefield image at 10X magnification to close-up detail simply by rotating the objective turret
  • Microscope is fully functional, independent of the camera
  • S71013 features a 3.0 MP live image
Warranty and Services
Lifetime limited warranty on microscope portion, excluding bulbs, camera and cords; Camera warranty: one year
Specifications
Magnification Power 10X to 30X
Objective Types Zoom
Product Type Digital Stereo Microscope
Head Style Binocular, Inclined/Rotating
Lighting Type Incidental/Transmitted
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov