Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisher Science Education™ Environmental Extremes Plant Growth Kit
SCI_ED

Catalog No. s02640 Shop All Fisherbrand Products
Change view
Click to view available options
Grade:
10 to 12
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Grade
S02640 10 to 12
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy

Choking hazard (1) Small parts. Not for children under 3 yrs.

Catalog No. S02640
This item is not returnable. View return policy

Choking hazard (1) Small parts. Not for children under 3 yrs.

Includes

  • Instructor manual with reproducible student data sheets
  • Stirring Rods
  • Weigh Dishes
  • Markers
  • Graduated Cylinders
  • Flasks
  • Gloves
  • Parafilm
  • Rulers
  • Cups
  • Deli Containers
  • Potting Soil
  • Petri Dishes
  • Seeds
  • Acid pH Solution Set
  • Sodium Chloride
  • Construction Paper
  • Masking Tape
  • Paper Towel Roll
  • Forceps
  • Magnifiers
  • Spoons
  • Write-N-Wipe Boards
  • Spray Bottle, 16 oz.
  • Alcohol Wipe, 70%
  • pH Meter
  • Digital Pocket Pen
  • Digital Scale, 2000Gx 0.1G
  • Stainless Steel scoop

Kit is designed to simulate conditions of acid rain, saline soils, drought and erosion in classroom.

  • Complete Curriculum Unit: Designed for a 4 to 6 week introduction to environmental studies
  • Hands-On Learning: Students simulate acid rain, saline soils, drought, and erosion conditions
  • Six Experimental Protocols: Teaches scientific method, data collection, and analysis using a plant science model
  • Custom Experiment Design: Encourages students to design their own experiments
  • Classroom Size: Supports 24 students
  • Reusable Components: Most kit materials are durable and reusable for future use
  • Educational Levels: Appropriate for grades 6 through 12
  • Comprehensive Guides: Includes reproducible student protocols and a detailed instructor manual
  • Flexible Configuration: Extended kits with pH meters and digital scales available (sold separately)
  • Note: Acids not included

Order Info

  • Designed for 24 students. This complete kit includes reproducible student protocols and instructor manual. (Acids not included.)
  • Can also be made available as an extended kit which includes STEM-related equipment you may not have in your lab, such as pH meters, buffer calibration kit and digital scales; contact your Fisher Science Education Representative for details.

Specifications

Grade 10 to 12
Product Type Environmental Extremes Plant Growth Kit
For Use With (Application) Complete introductory unit on environmental studies
Subject Science
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.