Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisher Science Education™ Foam Cups
SCI_ED

Catalog No. s04195 Shop All Fisherbrand Products
Click to view available options
Capacity (Metric):
177.4 mL
236.5 mL
354.8 mL
This item is not returnable. View return policy

Catalog No. S04195

This item is not returnable. View return policy

Commonly used in the science classroom.

Specifications

Material Foam
Capacity (Metric) 177.4 mL
Capacity (English) 6 oz.
Quantity 1000 Pk.
Product Type Cup
For Use With (Application) Used in the Science Classroom
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.