Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Fisherbrand™ Hazard and NFR Identification Write-On Labels





Ideal for adding vital hazard information to surfaces of small bottles, cans, and other containers.
$33.10 - $68.80
Specifications
Format | Label |
---|---|
Product Type | Hazard Identification Label |
For Use With (Application) | Chemical hazard |
Catalog Number | Quantity | Height (English) | Height (Metric) | Width (English) | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Quantity | Height (English) | Height (Metric) | Width (English) | Price | Quantity & Availability | |||
S43188
![]() ![]() ![]() |
1000/Roll | 1.5 in. | 3.81 cm | 3.88 in. |
Case of 1000 for $68.80
|
|
|||
S475646
![]() ![]() ![]() |
100/Roll | 3 in. | 7.6 cm | 5 in. |
Case of 100 for $33.10
|
![]() |
|||
Description
Blank hazard classification diamond can be filled in with the appropriate ratings.
Large Labels
- Spaces for Chemical Name, Hazards and Precautions
- Check boxes for goggles, gloves, apron and fume hood
- Dimensions: 5W × 3 in.H (12.7 × 7.6cm)
- Spaces for Chemical Name, Owner and Date
- Dimensions: 3.88W × 1.5 in.H (9.9 × 3.8cm)
Specifications
Label | |
Hazard Identification Label | |
Chemical hazard |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title