Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisher Science Education™ Electrical Tape
SCI_ED

Catalog No. S04192 Shop All Fisherbrand Products
Click to view available options
:
Black; Electrical Tape

Catalog No. S04192

Only null left
Add to Cart
Edge
Add to Cart

Electrical tape commonly used in the science classroom.

  • Black Tape for Industrial Applications
  • 30 yd. x 0.75 in. (27.4 m x 1.9 cm)
  • Best used for electrical insulation
  • Rubber adhesive / UL listed

Specifications

Product Type Electrical Tape
Color Black
For Use With (Application) For science classroom
Dimensions (L x W) 30 yd. x 0.75 in. (27.4 m x 1.9 cm)
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.