Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Fisher Science Education™ Total Hardness Test Kit
Shop All Fisherbrand Products

Click to view available options
:
Total Hardness Test Kit
Description
Includes
Hardness buffer, 30mL; hardness indicator powder, 10g; hardness titrant, 60mL; plastic vial
A stream or lake's hardness reflects the geology of the catchment area and sometimes provides a measure of the influence of human activity in a watershed. For the most part, however, hardness is a measure of the calcium and magnesium that enter a stream through the weathering of rock.
- One drop equals 10ppm hardness as CaCO3
- Perform the test and compare the color change to the EndPoint ID™ photograph on the instructions
Specifications
Specifications
Product Type | Total Hardness Test Kit |
For Use With (Application) | For analysis of natural water using a titration test method |
Includes | 30ml hardness buffer, 10g hardness indicator powder, 60ml hardness titrant, plastic vial |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction