Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Fisherbrand™ accumet™ AB15+ Basic and BioBasic™ pH/mV/°C Meters
Shop All Fisherbrand Products
Click to view available options
Display Type:
LCD
Description
Includes
- Meter Only package includes meter, power supply, electrode support (13-637-671) and operation manual.
- Meter Kit package includes items above plus an accumet liquid-filled, glass body single-junction combination pH Ag/AgCl Electrode (13-620-285) and separate ATC probe (13-620-19).
- BioBasic Meter Kit package for the life science lab includes meter, power supply, manual, electrode support and accuTupH™ rugged bulb, liquid-filled, glass body double-junction combination pH Combination Electrode (13-620-183) and separate ATC probe (13-620-19).
- Note: Each package is also available with a Certificate of Traceability to NIST™ Standards.
- Intuitive operation and improved user interface Large LCD guides user with plain-language prompts and error messages
- Display indicators keep you apprised of menu choice, measuring units, buffers standardized (in pH mode) and temperature
- No cryptic icons to learn: prompts and status/error messages are displayed in plain language
- Store up to 100 datapoints in nonvolatile memory
- Selectable resolution to 0.1, 0.01 or 0.001pH
- Five-button, soft-key input simplifies data entry
- Internal diagnostics
- Automatic recognition of 15 buffers and five-point calibration
- BNC and reference pin inputs, input for accuFet™ Electrode, ATC probe input, plus bidirectional RS-232 port
Order Info
Dimensions: 7.5L x 5.5W x 3.25 in.H (19 x 14 x 8cm). Net weight: 1.875 lb. (0.8kg).
Warranty and Services
accumet Model AB15+ is warranted to the end user to be free from defects in materials and workmanship under normal use for a period of three years.

Specifications
Specifications
Includes | Power supply, electrode support and accuTupH rugged bulb, double-junction combination pH electrode (13-620-183), separate ATC probe |
For Use With (Application) | pH, mV, Temperature |
Display Type | LCD |
Accuracy | Multiple |
Resolution | Multiple |
Detection Range | Multiple |
Electrical Requirements | 120V, 50/60Hz |
Voltage | 120 V |
Frequency | 60 Hz |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction