Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisherbrand™ accumet™ XL600 Benchtop Dual pH/ISE, Conductivity and Dissolved Oxygen Meters

Catalog No. 13636XL600 Shop All Fisherbrand Products
Click to view available options
Product Type:
Meter with Deluxe Kit
Meter with Standard Kit
Meter, Standard Kit, NIST Cert.
Multiparameter Meter

Includes

accumet XL600 dual pH/ISE, conductivity & DO benchtop meter, TRIS-compatible double junction refillable glass-body pH electrode (13-620-183A), ATC probe (13-620-19), 2-cell conductivity/temperature probe (13-620-100), electrode stand, RS232 and USB cables, 100/240V universal power supply

Provide advanced measurements with simple operation and offer a large, vivid color touch-screen display. Fisherbrand accumet XL600 pH/ISE/conductivity/DO benchtop meters offer advanced features with simple operation to help ensure your measurements are reliable and your laboratory processes are efficient.

  • High resolution color display shows data clearly from wide viewing angles and distances
  • Optimized touch-screen with glass/glass sensitivity provides great ease of use
  • Onscreen info includes date and time, stability indicator, alarms and context-specific help screens
  • One to six point pH calibration with 0.001 pH resolution
  • Automatic buffer recognition with six pH buffer sets or custom buffer option
  • Two to six point ISE calibration with 0.001 ppm, mg/L or molar resolution
  • ISE methods for known addition, known subtraction, analyte addition and analyte subtraction
  • Conductivity, TDS (total dissolved solids), salinity and resistivity modes
  • Dissolved oxygen mode calculates five-day BOD testing, OUR and SOUR
  • Salinity correction and barometric pressure compensation with built-in barometer to enhance DO accuracy
  • Display up to four measurements simultaneously (two pH or ISE, one conductivity, one DO)
  • Automatic or manual temperature compensation, displayed in °C or °F
  • Up to 10 password protected user ID accounts, each retains unique calibration and instrument settings with memory to store 2000 data points
  • RS232 and USB interfaces for printer or computer interfacing and real-time onscreen graphing of measurements
  • RJ45 Ethernet/Internet port to connect Local Area Network (LAN) or Internet
  • Bidirectional communication for LIMS connectivity and 3rd party software
  • Connectivity for peripherals such as keyboard, mouse or other USB device
  • Multilanguage interface with English, German, Chinese, French, Italian and Spanish
  • Two ports for optional stirring probe (Cat. No. 13-620-BSP) with five speed settings
  • Option to wall-mount meter to create additional bench space

NIST-Traceable Calibration Models (-CERT)

  • Help meet ISO, FDA, USDA and EPA quality assurance guidelines
  • Includes meter accuracy at five test points across pH range
  • Includes meter accuracy at five test points across corresponding mV range
  • Testing performed by accredited laboratory, includes NIST-Traceable certificate

Specifications

  • pH Range: -2.000 to 20.000 pH
  • pH Resolution: 0.1, 0.01, 0.001 pH
  • pH Accuracy: ±0.002 pH
  • ISE Range: 0.001 to 19999 ppm, mg/L or molar
  • ISE Resolution: 0.001, 0.01, 0.1, 1 (automatic)
  • ISE Accuracy: 0.5 % full scale (monovalent ion); 1 % full scale (divalent ion)
  • mV Range: ±2000.0mV
  • mV Resolution: 0.1mV
  • mV Accuracy: ±0.2mV or ±0.05 %, whichever is greater
  • mV Offset Adjustment: Up to ±150mV
  • Conductivity Range: 0.00μS to 500.0mS
  • Conductivity Resolution: 0.01, 0.1μS; 0.001, 0.01, 0.1mS
  • Conductivity Accuracy: ±1 % full scale
  • TDS Range: 0.00ppm to 500 ppt (TDS factor 1.00)
  • TDS Resolution: 0.01, 0.1 ppm; 0.001, 0.01, 0.1 ppt
  • TDS Accuracy: ±1 % full scale
  • Salinity Range: 0 to 80.0 ppt
  • Salinity Resolution: 0.01, 0.1 ppm; 0.001, 0.01, 0.1 ppt
  • Salinity Accuracy: ±1 % full scale
  • Resistivity Range: 2.000 Ohm to 20.0 MOhm
  • Resistivity Resolution: 0.01, 0.1 Ohm; 0.001, 0.1 kOhm; 0.01 MOhm
  • Resistivity Accuracy: ±1 % full scale
  • Dissolved Oxygen Range: 0 to 50mg/L; 0 to 600 % saturation
  • Dissolved Oxygen Range using 13-620-SSP: 0 to 20mg/L; 0 to 200 % saturation
  • Dissolved Oxygen Resolution: 0.01, 0.1mg/L; 0.1, 1 % saturation
  • Dissolved Oxygen Accuracy: ±0.1mg/L ±1 LSD, 0.1 % saturation
  • Barometric Pressure Range: 450 to 825mm Hg
  • Barometric Pressure Accuracy: ±1mm Hg + 1 LSD
  • Temperature Range: -5° to +105°C (23° to 221.0°F)
  • Temperature Resolution: 0.1°C (0.1°F)
  • Temperature Accuracy: ±0.2°C (±0.3°F)
  • Calibration Points: 1 to 6 points with automatic buffer recognition (ABR)
  • ISE Calibration Points: 2 to 6 points
  • ATC Temperature Calibration: Offset in 0.1°C increments; Offset range: ±5°C (±9°F)
  • Memory: 2000 data sets per user ID with date/time stamp
  • Self-Test: Yes
  • Inputs: Two BNC, two mini-phone ATC, 8 Pin DIN (conductivity), 8 Pin DIN (DO), ground
  • Output: RJ45, RS232, two USB, two stirring probe
  • Power Requirements: 100 to 240VAC, 50/60Hz
  • Dimensions (L x W x H): 24.6 x 17.3 x 9.9cm (9.7 x 6.8 x 3.9 in.)
  • Weight: 1.13kg (2.5 lb.)

Applications
Pharmaceutical, biotech, research, QA/QC, industrial, food, beverage

Certifications

CE, TUV 3-1, FCC Class A

Warranty and Services

3 years on meter

TRUSTED_SUSTAINABILITY

Specifications

Outputs RS232, USB-A, Mini-B USB, RJ45, stirrer port (2)
Accuracy Multiple
Accuracy (Conductivity) ±1% FS
Accuracy (DO) 0.1 +1 LSD
Accuracy (ISE) 0.5% full scale (monovalent ion), 1% full scale (divalent ion)
Accuracy (mV) ±0.2 mV
Accuracy (pH) ±0.002
Accuracy (Resistivity) ± 1% full scale
Accuracy (Salinity) ± 1% full scale
Accuracy (TDS) ± 1% full scale
Calibration Points (Conductivity) Automatic (4 points), maximum 1/range, manual (5 points), maximum 1/range
Calibration Points (ISE) 2 to 6
Calibration Points (pH) Up to 6 preset or 5 custom
Cell Constant 0.01 to 10
Certifications/Compliance NIST, ISO, FDA, USDA, EPA
Data Management Details 3 to 86400 sec.
Data Points 2000
Detection Range Multiple
Channels 2
Display Type LCD
Measurement Modes pH, mV, ISE, Ion, Concentration, Conductivity, TDS, Salinity, Resistivity, Temperature
Range (Conductivity) 0 to 500 mS
Range (mV) ±2000 mV
Range (pH) -2 to 20
Range (Resistivity) 3 W to 100 MW over 4 ranges: 0 to 20 kW, 20 to 200 kW, 200 kW to 2 MW, 2 to 100 MW
Range (Salinity) 0 to 80 ppt
Range (TDS) 0.050 ppm to 500 ppt (at TDS factor 1.00)
Resolution (ISE) 2/3/4 Digits
Resolution (pH) 0.1/0.01/0.001
Temperature Range (Metric) -5°C to +105°C
Includes Meter, TRIS compatible accuTupH pH electrode (13-620-183A), ATC probe (13-620-19), Conductivity/Temp. probe (13-620-100), electrode arm, RS-232 and USB cables, and 110/220V power supply
Inputs 2 BNC, 2 ATC
Model XL600 Dual pH/ISE/Conductivity/DO Kit
Product Type Meter with Standard Kit
Range (DO) 0 to 20mg/L (0 to 1272mbar), 0 to 200% saturation with 13-620-SSP electrode
Range (ISE) ±2000 mV
Resolution Multiple
Resolution (Conductivity) 0.001/0.01/0.1mS
Resolution (DO) 0.01mg/L (0.1mbar), 0.1% saturation
Resolution (mV) 0.1mV
Resolution (Resistivity) 0.01/0.1W, 0.001/0.1kW, 0.01MW
Resolution (Salinity) 0.01/0.1ppm, 0.001/0.01/0.1ppt
Resolution (TDS) 0.01/0.1ppm, 0.001/0.01/0.1ppt
Temperature Accuracy ±0.2°C (±0.3°F)
Temperature Resolution 0.1°C
Warranty 3 Years
Dimensions (L x W x H) 24.6 x 17.3 x 9.9cm (9.7 x 6.8 x 3.9 in.)
Length (English) 9.68 in.
Length (Metric) 24.6 cm
Width (Metric) 17.3 cm
Height (English) 3.89 in.
Height (Metric) 9.9 cm
Weight (English) 2.5 lb.
Weight (Metric) 1.13 kg
Voltage 100/240 V
Frequency 50/60Hz
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.