Learn More
Fisherbrand™ accumet™ XL600 Benchtop Dual pH/ISE, Conductivity and Dissolved Oxygen Meters
Provide advanced measurements with simple operation and offer a large, vivid color touch-screen display. Fisherbrand accumet XL600 pH/ISE/conductivity/DO benchtop meters offer advanced features with simple operation to help ensure your measurements are reliable and your laboratory processes are efficient.
Includes: accumet XL600 dual pH/ISE, conductivity & DO benchtop meter, electrode stand, RS232 and USB cables, 100/240V universal power supply
Description
Provide advanced measurements with simple operation and offer a large, vivid color touch-screen display. Fisherbrand accumet XL600 pH/ISE/conductivity/DO benchtop meters offer advanced features with simple operation to help ensure your measurements are reliable and your laboratory processes are efficient.
- High resolution color display shows data clearly from wide viewing angles and distances
- Optimized touch-screen with glass/glass sensitivity provides great ease of use
- Onscreen info includes date and time, stability indicator, alarms and context-specific “help” screens
- One to six point pH calibration with 0.001 pH resolution
- Automatic buffer recognition with six pH buffer sets or custom buffer option
- Two to six point ISE calibration with 0.001 ppm, mg/L or molar resolution
- ISE methods for known addition, known subtraction, analyte addition and analyte subtraction
- Conductivity, TDS (total dissolved solids), salinity and resistivity modes
- Dissolved oxygen mode calculates five-day BOD testing, OUR and SOUR
- Salinity correction and barometric pressure compensation with built-in barometer to enhance DO accuracy
- Display up to four measurements simultaneously (two pH or ISE, one conductivity, one DO)
- Automatic or manual temperature compensation, displayed in °C or °F
- Up to 10 password protected user ID accounts, each retains unique calibration and instrument settings with memory to store 2000 data points
- RS232 and USB interfaces for printer or computer interfacing and real-time onscreen graphing of measurements
- RJ45 Ethernet/Internet port to connect Local Area Network (LAN) or Internet
- Bidirectional communication for LIMS connectivity and 3rd party software
- Connectivity for peripherals such as keyboard, mouse or other USB device
- Multilanguage interface with English, German, Chinese, French, Italian and Spanish
- Two ports for optional stirring probe (Cat. No. 13-620-BSP) with five speed settings
- Option to wall-mount meter to create additional bench space
- Help meet ISO, FDA, USDA and EPA quality assurance guidelines
- Includes meter accuracy at five test points across pH range
- Includes meter accuracy at five test points across corresponding mV range
- Testing performed by accredited laboratory, includes NIST-Traceable certificate
- pH Range: -2.000 to 20.000 pH
- pH Resolution: 0.1, 0.01, 0.001 pH
- pH Accuracy: ±0.002 pH
- ISE Range: 0.001 to 19999 ppm, mg/L or molar
- ISE Resolution: 0.001, 0.01, 0.1, 1 (automatic)
- ISE Accuracy: 0.5 % full scale (monovalent ion); 1 % full scale (divalent ion)
- mV Range: ±2000.0mV
- mV Resolution: 0.1mV
- mV Accuracy: ±0.2mV or ±0.05 %, whichever is greater
- mV Offset Adjustment: Up to ±150mV
- Conductivity Range: 0.00μS to 500.0mS
- Conductivity Resolution: 0.01, 0.1μS; 0.001, 0.01, 0.1mS
- Conductivity Accuracy: ±1 % full scale
- TDS Range: 0.00ppm to 500 ppt (TDS factor 1.00)
- TDS Resolution: 0.01, 0.1 ppm; 0.001, 0.01, 0.1 ppt
- TDS Accuracy: ±1 % full scale
- Salinity Range: 0 to 80.0 ppt
- Salinity Resolution: 0.01, 0.1 ppm; 0.001, 0.01, 0.1 ppt
- Salinity Accuracy: ±1 % full scale
- Resistivity Range: 2.000 Ohm to 20.0 MOhm
- Resistivity Resolution: 0.01, 0.1 Ohm; 0.001, 0.1 kOhm; 0.01 MOhm
- Resistivity Accuracy: ±1 % full scale
- Dissolved Oxygen Range: 0 to 50mg/L; 0 to 600 % saturation
- Dissolved Oxygen Range using 13-620-SSP: 0 to 20mg/L; 0 to 200 % saturation
- Dissolved Oxygen Resolution: 0.01, 0.1mg/L; 0.1, 1 % saturation
- Dissolved Oxygen Accuracy: ±0.1mg/L ±1 LSD, 0.1 % saturation
- Barometric Pressure Range: 450 to 825mm Hg
- Barometric Pressure Accuracy: ±1mm Hg + 1 LSD
- Temperature Range: -5° to +105°C (23° to 221.0°F)
- Temperature Resolution: 0.1°C (0.1°F)
- Temperature Accuracy: ±0.2°C (±0.3°F)
- Calibration Points: 1 to 6 points with automatic buffer recognition (ABR)
- ISE Calibration Points: 2 to 6 points
- ATC Temperature Calibration: Offset in 0.1°C increments; Offset range: ±5°C (±9°F)
- Memory: 2000 data sets per user ID with date/time stamp
- Self-Test: Yes
- Inputs: Two BNC, two mini-phone ATC, 8 Pin DIN (conductivity), 8 Pin DIN (DO), ground
- Output: RJ45, RS232, two USB, two stirring probe
- Power Requirements: 100 to 240VAC, 50/60Hz
- Dimensions (L x W x H): 24.6 x 17.3 x 9.9cm (9.7 x 6.8 x 3.9 in.)
- Weight: 1.13kg (2.5 lb.)
Certifications
CE, TUV 3-1, FCC Class A
Warranty and Services
3 years on meter
Specifications
RS232, USB-A, Mini-B USB, RJ45, stirrer port (2) | |
±1% FS | |
0.5% full scale (monovalent ion), 1% full scale (divalent ion) | |
±0.002 | |
± 1% full scale | |
Automatic (4 points), maximum 1/range, manual (5 points), maximum 1/range | |
Up to 6 preset or 5 custom | |
NIST, ISO, FDA, USDA, EPA | |
2000 | |
2 | |
pH, mV, ISE, Ion, Concentration, Conductivity, TDS, Salinity, Resistivity, Temperature | |
±2000 mV | |
3 W to 100 MW over 4 ranges: 0 to 20 kW, 20 to 200 kW, 200 kW to 2 MW, 2 to 100 MW | |
0.050 ppm to 500 ppt (at TDS factor 1.00) | |
0.1/0.01/0.001 | |
Meter, electrode arm, RS-232 and USB cables, and 110/220V power supply | |
XL600 Dual pH/ISE/Conductivity/DO Meter | |
±2000 mV | |
0.001/0.01/0.1mS | |
0.1mV | |
0.01/0.1ppm, 0.001/0.01/0.1ppt | |
±0.3°C | |
3 Years | |
9.68 in. | |
17.3 cm | |
9.9 cm | |
1.13 kg | |
50/60Hz |
Multiple | |
0.1 +1 LSD | |
±0.2 mV | |
± 1% full scale | |
± 1% full scale | |
2 to 6 | |
0.01 to 10 | |
3 to 86400 sec. | |
Multiple | |
LCD | |
0 to 500 mS | |
-2 to 20 | |
0 to 80 ppt | |
2/3/4 Digits | |
-5°C to +105°C | |
2 BNC, 2 ATC | |
0 to 20mg/L (0 to 1272mbar), 0 to 200% saturation with 13-620-SSP electrode | |
Multiple | |
0.01mg/L (0.1mbar), 0.1% saturation | |
0.01/0.1W, 0.001/0.1kW, 0.01MW | |
0.01/0.1ppm, 0.001/0.01/0.1ppt | |
0.1°C | |
24.6 x 17.3 x 9.9cm (9.7 x 6.8 x 3.9 in.) | |
24.6 cm | |
3.89 in. | |
2.5 lb. | |
100/240V |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.