Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Boekel Scientific™ Dricycler
GSA_VAGREENER_CHOICESDP

Catalog No. p-4432598 Shop All Fisherbrand Products
Change view
Click to view available options
Size:
Small
Large
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
08-645-33 Large
08-645-32 Small
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 08-645-33 Supplier Boekel Scientific™ Supplier No. 134441
Only null left
Add to Cart
Add to Cart
Request a Quote

The Dricycler with automatic desiccant regenerates the silica gel desiccant every five hours. Ideal for storing crucibles, storing reagents, and drying specimens.

  • Automatically regenerates desiccant every five hours
  • Adjustable shelves
  • Supplied with 2 shelves
  • Hand crafted from stainless steel and glass

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Specifications

Cabinet Style Front-loading
Material Stainless Steel, Glass
Product Type Dricycler
For Use With (Application) Storing reagents or soil samples, drying biological specimens, storage sludge samples, cooling down from 40° to 150°C.
Includes Adjustable shelves, neoprene door gasket
Shelf Positions 1/2 in. Increments
Size Large
Warranty 2 Years
Dimensions (L x W x H) 23 x 20.5 x 22.25 in. (58.4 x 52.1 x 57.2 cm)
Length (English) 23 in.
Length (English) Chamber 19.75 in.
Length (Metric) 58.4 cm
Length (Metric) Chamber 50.2 cm
Width (English) 20.5 in.
Width (English) Chamber 15.75 in.
Width (Metric) 52.1 cm
Width (Metric) Chamber 40 cm
Height (English) 22.25 in.
Height (English) Chamber 21.75 in.
Height (Metric) 57.2 cm
Height (Metric) Chamber 55.2 cm
Weight (English) 62 lb.
Weight (Metric) 28.1 kg
Voltage 115 VAC
Wattage 35 W
Frequency 50/60 Hz
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.