Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Boekel Scientific™ Dricyclers Available on GSA/VA Contract for Federal Government customers only.

Dricycler for storing crucibles, storing reagents and drying specimens.

Manufacturer:  Boekel Scientific™ 134441

 View more versions of this product

Catalog No. 08-645-33


Add to cart

Includes: 2 stainless steel shelves

Description

Description

  • Automatically regenerates desiccant every five hours
  • Adjustable shelves

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Warranty and Services

Two years

Specifications

Specifications

23.0 x 20.5 x 21.7 in. (58.4 x 52.1 x 55.2cm)
2 Years
58.4cm
Dricycler
Storing reagents or soil samples, drying biological specimens, storage sludge samples, cooling down from 40° to 150°C
55.2cm
50.2cm
35w
28.1kg
115V 50/60Hz, 35W
57.2cm
Stainless Steel and Glass
52.1cm
50/60Hz
Adjustable shelves, neoprene door gasket
115VAC
62 lb.
40cm
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit