Learn More
Fisherbrand™ Isotemp™ Digital Dry Baths/Block Heaters
Shop All Fisherbrand ProductsDescription
Requires
Unit must be ordered with block(s)
Fisherbrand™ Isotemp Digital Dry Baths/Block Heaters offer a range of configurations with interchangeable modular blocks for a variety of application needs.
Fisherbrand™ Isotemp Digital Dry Baths/Block Heaters offer a range of configurations with interchangeable modular blocks for a variety of application needs. Designed for basic to advanced applications with a maximum temperature of 130°C, a timer and available in 1, 2 and 4 block sizes.
- Digital controls and display of time and temperature
- Advanced internal temperature sensing probe for outstanding temperature accuracy and control
- Precise temperature control with PID circuit
- Temperature calibration enables to offset the temperature to desired value
- Timer allows the user to accurately monitor the heating time
- Wide range of interchangeable aluminum alloy heat blocks provide versatility and allow for easy cleaning and disinfecting
- Powder-coated steel body construction ensures durability
- Built-in over-temperature protection device warrants sample and user safety
- Temperature deviation adjustment
Applications:
Sample preparation, enzyme preservation, enzyme-substrate reactions, DNA amplification, electrophoresis gel degeneration and serum coagulation
Warranty and Services
2-year Thermo Fisher Scientific standard, limited warranty. Contact your sales representative for details.
Specifications
Specifications
| Certifications/Compliance | CE, cULus, RoHS |
| Controller Type | PID |
| Material | Powder-coated Steel |
| No. of Places | 1 |
| Plug Type | CN, EU, UK |
| Relative Humidity | <80% |
| Temperature Range (Metric) | Ambient +5° to 130°C |
| Type | Dry Bath Block |
| Fuse | 250V 2.5A |
| Heat Up Time | ≤ 20 min. 30° to 130°C |
| Show More |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.