Learn More
Fisherbrand™ Model 705 Sonic Dismembrator Accessory
4 Tip horn for use with 500 or 700 watt sonicator systems; replacement tip also available.
$371.00 - $2370.00
Requires: Cat. No. FB4659
Catalog Number | Product Type | For Use With (Equipment) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Product Type | For Use With (Equipment) | Price | Quantity & Availability | |||||
15-338-322
![]() |
Replacement Tip | 4-Tip Horn ( #4659) |
Each for $371.00
|
|
|||||
15-338-319
![]() |
4-tip Horn | FB705 Sonic Dismembrator and Q700 Sonicator |
Each for $2,370.00
|
|
|||||
Description
- For processing 0.5 to 15ml volumes
- Process 4 samples at one time