Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisherbrand™ Refrigerator/Freezer Thermometer
SCI_ED

Catalog No. S66277 Shop All Fisherbrand Products
Change view
Click to view available options
Accuracy:
±0.5°C
±1°C
Resolution:
0.1°C
1°C
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Accuracy Resolution
S66277 ±1°C 1°C
S66278 ±0.5°C 0.1°C
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S66277
Add to Cart
Edge
Add to Cart

Insulated sensors protect against rapid temperature changes. Fisherbrand Refrigerator/Freezer Thermometers accurately monitor temperatures in freezers, water baths, heating blocks, incubators and refrigerators with bottle-enclosed temperature buffered sensor.

  • Nontoxic glycol solution in bottle insulates sensor from rapid temperature changes when door is open
  • Triple display shows high, low and current temperatures, and min/max feature monitors high and low readings for any time period
  • Alarm feature provides an alert when temperature rises above or falls below a setpoint
  • 10 ft. micro-cable permits refrigerator doors to close on it
  • Velcro™ fasteners and magnetic strips are provided for mounting of both thermometer and bottle sensor
  • Supplied with one-year AA battery

S66277

  • Range: -50° to +70°C (-58° to +158°F)
  • Resolution: 1°
  • Accuracy: ±1°C

S66278

  • High-resolution, high-accuracy and min/max memory
  • Range: -50° to +70°C (-58° to +158°F)
  • Resolution: 0.1°
  • Accuracy: ±0.5°C

Certifications

Individually serial-numbered Traceable™ Certificate is provided from an ISO 17025 calibration laboratory accredited by A2LA™. It indicates traceability to standards provided by NIST™.

Specifications

Certifications/Compliance Individually serial-numbered Traceable™ Certificate is provided from an ISO 17025 calibration laboratory accredited by A2LA™. It indicates traceability to standards provided by NIST™.
Dimensions (L x W x H) 4.25 x 2.75 in. (10.7 x 6.9 cm)
For Use With (Application) Monitoring liquids, air/gas or semi solids in freezers, water baths, outdoor tanks, incubators and refrigerators
Includes AAA battery, Velcro™ and magnetic strips are provided for mounting of both thermometer and bottle sensor
Length (English) Overall 4.25 in.
Length (Metric) 21 cm
Length (Metric) Overall 10.7 cm
Material Plastic
Resolution 1°C
Weight (English) 4 oz.
Weight (Metric) 110 g
Width (English) 2.75 in.
Product Type Fridge/Freezer Digital Thermometer
Max. Temperature (English) 158°F
Max. Temperature (Metric) 70°C
Min. Temperature (English) -58°F
Min. Temperature (Metric) -50°C
Temperature Range (English) -58°F to +158°F
Temperature Range (Metric) -50°C to +70°C
Calibrated Yes
For Use With (Equipment) Refrigerators/Freezers
Accuracy ±1°C
Display Digital LCD
Width (Metric) 6.9 cm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.