Learn More
Fisherbrand™ Traceable™ Decimal Stopwatch
Shop All Fisherbrand ProductsDescription
Includes
Stopwatch, lanyard
Precise, Traceable timing for enzyme tests, viscosity measurements, animal studies, and all timed lab tests. Fisherbrand Traceable Decimal Stopwatch is tough with water-resistant ABS plastic case which seals out moisture, dust, and fumes encountered in harsh lab, plant, or field environments.
Made for scientists in industrial, educational, hospital, and governmental labs who insist on the finest instrumentation. Engineered for ease of operation, accuracy, and reliability.
- Quartz-crystal design is accurate to 0.0005% — over 100 times more accurate than other stopwatches
- Decimal stopwatch's four timing formats/resolution are: hr./min./sec.; 1/100 sec.; 1/1000 min; 1/100,000 hr.
- Maximum readouts are 19.999999 hours, 1199.999 minutes, 71999.99 seconds, or 19 hr./59 min./59 sec./99 hundredths
- Stopwatch functions include single-action, time-out, interval-split, cumulative split, and continuous roll-over timing
- Triple display shows cumulative splits, interval splits, and running time
- 500 recallable memories display the lap, split and running time
- Memory feature also displays the fastest, slowest, and average lap time
- Unit displays time of day, date, day of the week and features a time-of-day alarm
- Additional features include countdown alarm, countdown then countup alarm, countdown repeat, pacer, and stroke functions
- Timing range is 19 hours, 59 minutes, 59 seconds, and 99 hundredths
- Accuracy: 0.0005%
- Size: 6.4 x 7.6 x 1.9 cm (2.5 x 3 x 0.75 in.)
- Weight: 2.5 oz.
Order Info
Replacement battery: Cat. No. 02-440-1A
Certifications
Individually serial-numbered Traceable™ Certificate is provided from an ISO 17025 calibration laboratory accredited by A2LA. It indicates traceability to standards provided by NIST.
Specifications
Specifications
Product Type | Digital Stopwatch |
Time Range | 10 hr. |
Alarms | Audible |
Accuracy | 5e-06 |
Resolution | 0.01 sec. |
Display | LCD |
Battery Type | Lithium Cell, 3V (CR2032) |
Dimensions (L x W x H) | 2.5 x 3 x 0.75 in. (6.35 x 7.62 x 1.9 cm) |
Length (English) | 2.5 in. |
Length (Metric) | 6.35 cm |
Show More |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.