Learn More
Fisherbrand™ Traceable™ Humidity/Temperature Clipboard with Scientific Calculator and Stopwatch
Catalog No. 1166117 Shop All Fisherbrand Products
Includes
Supplied with two-year silver-oxide batteries.
Four devices in one convenient clipboard design.
  • Removable humidity/temperature pen (also available separately, 11-661-14) updates readings every second
  • Pen continuously and simultaneously displays both relative humidity and °C/°F
  • Pen memory saves and displays minimum and maximum readings during any user-defined time period
  • Clip securely holds papers
  • Hole allows unit to be hung on wall
Hygrometer. Range of 2.5 to 98% with an accuracy of ±3.5% at the midrange and ±5.5% at the ends of the range. Resolution of 1% RH.
Thermometer. Range of 0° to 50°C (32° to 122°F) with an accuracy of ±1°C (1.8°F) across the full range and a resolution of 0.1°C.
Digital Stopwatch with Alarm. Displays date, time of day, and alarm setting. Times to 24 hours. Resolution is 0.01 sec. for the first 30 minutes, and 1 second thereafter. Accuracy is ±0.01%.
Scientific Calculator. Performs most lab and plant calculations. Over 50 functions, including log, sin, cos, tan, pi, mean, hex, degrees, square root, statistics, exponents, arithmetic, and parenthesis.
Order Info
Dimensions: 13L x 9W x 1/8 in.H (34.29 x 22.86 x 3cm). Weight: 11 oz. (311g).
Certifications
An individually serial-numbered ISO 17025 certificate is included to indicate instrument traceability to standards provided by NIST.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.