Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisherbrand™ Traceable™ Flip-Stick™ Thermometer

Catalog No. p-4418227 Shop All Fisherbrand Products
Change view
Click to view available options
Accuracy:
±0.3°C
±1°C
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Accuracy
06-664-26 ±0.3°C
14-648-45 ±1°C
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 06-664-26
Only null left
Add to Cart
Add to Cart

Multipurpose, fold-out design makes taking probe readings in any position a snap

Fisher Scientific™ Traceable™ Flip-Stick™ Thermometer with fold-out design makes taking probe readings in any position a snap.

  • Features °F/°C switch
  • Stainless-steel probe
  • Shockproof and drop-proof up to 5 ft.
  • Max/min temperature reading over any period
  • Wide temperature range cover most routine measurements
  • Position probe at any angle
  • Protect probe in fold-up position
  • 0.75 in. Display is easy to read
  • Rapid reading update—every second
  • HOLD switch freezes display to capture a temperature

Certifications

Individually serial-numbered Traceable Certificate is provided from ISO 17025 calibration laboratory accredited by A2LA indicating traceability to standards provided by NIST (National Institute of Standards and Technology).

Specifications

Certifications/Compliance ISO 17025, A2LA, NIST
Dimensions (L x W x H) 11 x 0.6 in. (28 x 1.5 cm)
For Use With (Application) Food, gases, samples, reagents, water baths and semi solids
Includes Stainless Steel probe
Length (English) Probe 4.5 in.
Length (Metric) 280 mm
Length (Metric) Probe 11.4 cm
Material Plastic
Resolution 0.1°C
Weight (English) 2.4 oz.
Weight (Metric) 68 g
Width (English) 0.6 in.
Product Type Digital Thermometer
Length (English) 11 in.
Max. Temperature (English) 572°F
Max. Temperature (Metric) 300°C
Min. Temperature (English) -58°F
Min. Temperature (Metric) -50°C
Temperature Range (English) -58°F to +572°F
Temperature Range (Metric) -50°C to +300°C
Calibrated Yes
Accuracy ±0.3°C
Display Digital
Width (Metric) 1.5 cm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.