Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisherbrand™ Traceable™ International Standards Extreme-Accuracy Digital Thermometer

Catalog No. 15078179 Shop All Fisherbrand Products
Change view
Click to view available options
Temperature Range (Metric):
-2.00°C to +2.00°C
-50°C to +70°C
23.00°C to 27.00°C
35°C to 39°C
Temperature Range (English):
-58° to +158°F
28.4°F to 35.6°F
37.00° F
73.4°F to 80.6°F
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Temperature Range (Metric) Temperature Range (English)
15-078-179 23.00°C to 27.00°C 73.4°F to 80.6°F
15-078-178 -2.00°C to +2.00°C 28.4°F to 35.6°F
15-078-180 35°C to 39°C 37.00° F
15-078-181 -50°C to +70°C -58° to +158°F
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 15-078-179
Only null left
Add to Cart
Add to Cart

Traceable™ International Standards Extreme-Accuracy Digital Thermometers ideal for measuring temperatures in water baths, cuvets, gas systems, reagents, standards solutions, ovens, incubators, pharmaceuticals, petroleum products, foods, and wastewater.

Units read 0.00, 25.00, 37.00, or a combination of all three in °F and °C.

  • Provides accurate readings with tip penetration of 0.33 in.
  • Records time/date for min/max readings and hi/lo alarm alerts
  • Displays minimum/maximum and current temperatures
  • Min/max monitors readings over any period; displays exact time/date of min/max temperature
  • Alarm provides two visual LEDs and two audio alerts when temperature exceeds set points; displays exact time/date of dual thermometer alarms
  • Programmable in 0.1° increments; alarms signal continuously even if temperature returns to non-alarm range
  • Resolution: 0.01°
  • Accuracy: ±0.05°C within ±2°C of standard
  • Probe Dia.: 0.12 in. (0.30 cm)
  • Stem Length: 6.25 in. (15.87 cm)
  • Overall Length: 9 in. (22.86 cm)
  • Cable: 5 ft. (1.5 m)

Certifications

Individually serial-numbered Traceable Certificate is provided from an ISO 17025 calibration laboratory accredited by A2LA™; indicates traceability to standards provided by NIST™.

Compliance

Meets ITS-90, International Temperature Standard.

Specifications

Certifications/Compliance ISO 17025, A2LA, NIST
Diameter (English) Probe 0.12 in.
Diameter (Metric) Probe 3 mm
Dimensions (L x W x H) 4.25 x 2.75 x 0.75 in. (10.7 x 6.9 x 1.9 cm)
For Use With (Application) Measuring temperatures in water baths, cuvets, gas systems, reagents, standards solutions, ovens, incubators, pharmaceuticals, petroleum products, foods and wastewater
Height (English) 0.75 in.
Height (Metric) 1.9 cm
Includes Stainless Steel probe, High-impact, chemical-resistant ABS plastic case, stand, Velcro™, magnetic strips, wall mount, two AA batteries (allow continuous, 1-year always-on monitoring)
Length (English) Probe 6.25 in.
Length (Metric) 107 mm
Length (Metric) Probe 15.9 cm
Material ABS Plastic
Resolution 0.01°C
Weight (English) 5 oz.
Weight (Metric) 140 g
Width (English) 2.75 in.
Product Type Digital Thermometer
Length (English) 4.25 in.
Max. Temperature (English) 80.6°F
Max. Temperature (Metric) 27°C
Min. Temperature (English) 73.4°F
Min. Temperature (Metric) 23°C
Temperature Range (English) 73.4°F to 80.6°F
Temperature Range (Metric) 23.00°C to 27.00°C
Calibrated Yes
For Use With (Equipment) Water Baths
Battery Type AA
Accuracy ±0.05°C
Display LED
Width (Metric) 6.9 cm
Scale Reads to 25.00
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.