Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisherbrand™ Traceable™ Jumbo Refrigerator/Freezer Thermometers

Catalog No. 14648232 Shop All Fisherbrand Products
Click to view available options
Probe Type:
Naked Sensor
Sealed Bottle Probe
This item is not returnable. View return policy
This item is not returnable. View return policy

Includes

Unit is supplied with a Traceable Certificate, flip-open stand for lab bench, slot for wall mounting, one AAA alkaline battery and Velcro™ and magnetic strips to attach to any surface

Perfect for monitoring freezers, water baths, heating blocks, incubators and refrigerators

Perfect for monitoring freezers, water baths, heating blocks, incubators and refrigerators. Fisher Scientific™ Traceable™ Jumbo Refrigerator/Freezer Thermometer accurately monitors temperature with an enclosed temperature-bufferedsensor.

  • Bottle model (14-649-223) insulates sealed sensor from rapid temperature changes when refrigerator door is opened
  • Thermometer fulfills all CDC thermometer and vaccine thermometer requirements
  • Easy-view, jumbo 1.5 in. (38.1 cm) high digits may be read from 30 ft. (9m)
  • Sensor and 10 ft. cable perform accurately even when both are under water
  • High-impact, chemical-resistant ABS plastic case
  • Triple display simultaneously shows maximum, minimum and current probe temperatures plus room temperature
  • Min./max. monitors high/low readings overnight, on weekends or for any time period — a significant advantage over 'current reading-only' glass thermometers
  • Range is -58° to +158°F and -50° to +70°C with a resolution of 0.01° and accuracy of ±1°C
  • Alarm feature provides alert when temperature rises above or falls below a setpoint
  • Alarm is programmable in 1° increments
  • 10 ft. (3m) micro-cable permits refrigerator doors to close on it
  • Solid-state probes eliminate mercury contamination in refrigerators
  • Sealed probe is housed in a miniature bottle filled with nontoxic glycol; solution is GRAS (Generally Recognized as Safe)
  • Eliminates concerns about incidental contact with food or drinking water

Order Info

  • Case: 3.87 x 4.37 x 0.87 in. (9.83 x 11.1 x 2.2cm)
  • Sealed bottle probe measures 1 x 2.5 in. (2.5 x 6.4cm)
  • Naked sensor is 0.18 dia. x 0.75 in. long (4.6 x 19mm)
  • Replacement battery (02-440-3A) available

Certifications

Individually serial-numbered Traceable Certificate is provided from an ISO 17025 calibration laboratory accredited by A2LA™; indicates traceability to standards provided by NIST™.

Specifications

Certifications/Compliance Individually serial-numbered Traceable Certificate is provided from an ISO 17025 calibration laboratory accredited by A2LA™; indicates traceability to +standards provided by NIST™.
Dimensions (L x W x H) 3.88 x 3.87 x 4.37 in. (9.8 x 9.82 x 11.1 cm)
For Use With (Application) Monitoring freezers, water baths, heating blocks, incubators and refrigerators
Height (English) 4.37 in.
Height (Metric) 11.1 cm
Includes Naked sensor, traceable certificate, flip open stand, wall mount, Velcro™, magnetic strips and AAA Alkaline battery
Length (English) Cable 120 in.
Length (Metric) 98 mm
Length (Metric) Cable 304.8 cm
Material ABS Plastic
Probe Type Naked Sensor
Resolution 0.1°C
Weight (English) 4 oz.
Weight (Metric) 110 g
Width (English) 3.87 in.
Product Type Digital Thermometer
Length (English) 3.88 in.
Max. Temperature (English) 158°F
Max. Temperature (Metric) 70°C
Min. Temperature (English) -58°F
Min. Temperature (Metric) -50°C
Temperature Range (English) -58°F to +158°F
Temperature Range (Metric) -50°C to +70°C
Calibrated Yes
Accuracy ±1°C
Display Digital
Width (Metric) 9.82 cm
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.