Learn More
Fisherbrand™ Traceable™ Tubing Sets
Shop All Fisherbrand Products
Click to view available options
For Use With:
Medium Flow Variable-Speed Pump
Medium/High Flow Variable-Speed Pump
Ultra Low Flow and Low Flow Variable-Speed Pump
Description
For use with Variable-Speed Pumps
- Replacement set of tubing and fittings/nipples
- Identical to the set supplied with pump
- Supplied with 1/50, 1/16, 3/32, 3/16 and 1/4 in. I.D. tubing and corresponding fittings/nipples
Specifications
Specifications
| Description | Tubing Replacemant Set |
| Diameter (English) Inner | 1/50, 1/16, 3/32, 3/16, 1/4 in. |
| Diameter (Metric) Inner | 0.508, 1.58, 2.38, 4.76, 6.35 mm |
| For Use With | Medium/High Flow Variable-Speed Pump |
| Product Type | Tubing Set |