Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Fisher Scientific™ Vehicle Power Adapter
Shop All Fisherbrand Products
Click to view available options
For Use With (Equipment):
Dry blocks
Description
Convient way to charge on the go.
- 12 volt
- Charging light
- Suitable length cord
- Shaped for grip to plug and unplug easily
Specifications
Specifications
| Color | Black |
| Description | AC adapter for Vehical charging |
| For Use With (Equipment) | Dry blocks |
| Plug Type | Car Adapter |
| Type | Vehicle Power Adapter |
| Includes | Charging light to easily confirm product is securely plugged in |
| Electrical Requirements | 12 V |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction