Fisherbrand™ 96-Well Plates Available on GSA/VA Contract for Federal Government customers only.

For EIA and other applications

 View more versions of this product

Catalog No. 21-377-203

  Check Availability
Add to Cart

    Specifications

    Lid Yes
    For Use With (Application) EIA and other applications
    Binding Property High
    Material Polystyrene
    No. of Wells 96.0
    Sterile No
    Type 96-Well Assay Microplate
    Certifications/Compliance Certified
    Volume (Metric) Well 450μL
    Well Design Rounded Square (C),Rounded Square (C)
    Closure Yes
    Color Clear
    Applications EIA and other applications
    Total Count 96.0
    Well Shape Round

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit