Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisherbrand™ Absorption Ultra Micro Rectangular Cells with PTFE Lid

Catalog No. p-4903607 Shop All Fisherbrand Products
Click to view available options
Capacity (Metric):
50 μL
100 μL
Description:
Center:15mm
Center:8.5mm
This item is not returnable. View return policy
This item is not returnable. View return policy

Includes

PTFE lid or stopper

Fit standard spectrophotometer cell holders

  • All cells are 12.5mm wide
  • 1, 5 or 10mm path length rectangular cells
  • Reduce reagent use with Ultramicro cells
  • Quartz windows for transmission from far ultraviolet through the infrared
  • Glass windows for routine work in ultraviolet region and visible region

Specifications

Type Ultra-micro Absorption Cell
Description Center:8.5mm
Capacity (Metric) 100 μL
Material Quartz
Closure Material PTFE
Width (Metric) Interior 2mm
Quantity Ea., 8 Ea./Cs.
Path Length 10 mm
Wavelength Range 200 to 2500 nm
Height (Metric) 45 mm
For Use With (Equipment) Standard Spectrophotometer Cell Holder
Width (Metric) 12.5 mm
For Use With (Application) Transmission from far ultraviolet through the infrared
Shape Rectangle
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.