Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Fisherbrand™ Acid/Alkali Test Papers
Shop All Fisherbrand Products
Click to view available options
pH Range:
<3: Blue, >5.2: Red
0 to 14
0 to 7
7 to 14
Type:
Blue Litmus Test Strip
Congo Red Test Strip
Litmus Neutral Test Strip
Litmus Red Test Strip
Description
- A foolproof pH test to determine if a solution is acidic
- The Blue Litmus test paper does not come with a color chart as it cannot provide you with a defined pH value
- The test strip will turn red if dipped into an acidic solution (pH <7)
- It will remain unchanged in an alkaline solution
- We have tested the blue litmus with multiple pH buffers
- The blue paper is blue at pH 8
- At pH 7, the sample is maybe just a little red
- The real change to red seems to start with pH 6
- 5 and lower
- Neutral Litmus test paper tests for Neutrality of a solution, or its acidity or alkalinity
- The test strip will turn red if dipped into an acidic solution (pH <7) or blue if dipped into an alkaline solution (pH >7)
- Red Litmus test paper is a foolproof pH test to determine if a solution is alkaline
- The test strip will turn blue if dipped into an alkaline solution (pH >7)
- Color chart not included
Order Info
100 Strips per vial
Specifications
Specifications
Certifications/Compliance | OSHA, 29 CFR 1910.133, EN166 |
pH Range | 0 to 7 |
Quantity | 24 x 100 Strips |
Type | Blue Litmus Test Strip |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction