Learn More
Alkacid™ Wide-Range Test Strips
Catalog No. a980 Shop All Fisherbrand Products
Click to view available options
pH Range:
2 to 10
  • Change to red (pH 2), orange (pH 4), yellow (pH 6), green (pH 8), blue (pH 10)
Order Info
In amber vials of 100 strips
TRUSTED_SUSTAINABILITY
Specifications
pH Range 2 to 10
For Use With (Application) Provide wide-range of five distinct color changes in pH range 2 through 10, color changes: red (pH 2), orange (pH 4), yellow (pH 6), green (pH 8), blue (pH 10)
Quantity 12 Pack
Type pH Test Strip
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.