Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisherbrand™ Amber Straight Sided Round Bottles without Cap

Catalog No. 02912298 Shop All Fisherbrand Products
Click to view available options
Capacity (Metric):
125 mL

Fisherbrand Amber Straight Sided Round Bottle are designed without shoulders for maximum storage capacity.

Amber Straight Sided Round Glass Jars are designed without shoulders for maximum storage capacity and UV protection. These multipurpose jars are very versatile. Our straight sided jars are available in a wide variety of sizes, from 1oz to 32oz.

  • Wide mouth for easy filling

Specifications

Capacity (Metric) 125 mL
Capacity (English) 4 oz.
Color Amber
Includes Only jar
Material Glass
Quantity 24/PK.
Closure Included No
Mouth Wide
Product Type Straight Sided Round Bottle
Closure Size 58-400 mm
Closure Type Screw Cap
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.