Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bio Plas™ Astral™ Inoculation Device System

Catalog No. p-2410978
Click to view available options
Product Type:
Astral Handle
Inoculating Loop
Streaking Needle
Color:
Black
Blue
White
Yellow
This item is not returnable. View return policy
This item is not returnable. View return policy

Includes

Autoclavable handle with every case

Economical, disposable inoculation system designed for safety and dependable sterility

Economical, disposable inoculation system designed for safety and dependable sterility

Calibrated using Evans Blue Dye spectrophotometric procedure and gravimetric method

  • Free of DNA, RNase, DNase, pyrogens and PCR inhibitors
  • Astral loops, needles, and Clone-Piks are packaged static free into individual tubular chambers, which are then hermetically sealed with aluminum film
  • A dust cover is heat-shrunk onto the unit before it is irradiation sterilized
  • Use on an individual basis, or for quantitative procedures such as sampling, urine counts, or serial dilutions

To use:

  • Puncture aluminum film with the handle, pushing down until it attaches to the loop, needle, or Clone-Pik
  • Withdraw handle; inoculation device is ready for many applications

Order Info

Supplied 10 racks of 96 to a case

Specifications

Disposable Single-use
Volume 1 μL
Sterility Sterile
Color Yellow
Product Type Inoculating Loop
For Use With (Equipment) Astral™ inoculation device system
Includes Autoclavable handle
Packaging Type Rack
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.