Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Fisherbrand™ Autoclavable Biohazard Waste Bags
Shop All Fisherbrand Products

Click to view available options
Capacity (English):
13 to 16 gal.
20 to 30 gal.
44 gal.
7 to 10 gal.
8 to 12 gal.
Dimensions (L x W):
23 x 19 in.
30 x 24 in.
35 x 25 in.
38 x 31 in.
48 x 37 in.
Description
Dual tested compliant
Autoclaving is a very common way to sterilize infectious waste through the use of high temperature, high pressure steam over a specific time period. Sterilized waste is less expensive to dispose. Dual-tested autoclave bags are ASTM D1922 Elmendorf Tear tested to exceed 480g of tear strength (MD and TD) and ASTM D1709 tested to exceed 165g dart drop strength.
- Integral steam processing indicator that turns brown when autoclaved
- Bags comply with US DOT 49CFR Sections 173-197 and specific state regulations
- Operating temperature tolerance up to 285degreeFdegreeAvailable in various sizes
- Bag with black print
- Gauge: 2mil
Specifications
Specifications
Capacity (English) | 7 to 10 gal. |
Color | Red, Black Print |
Dimensions (L x W) | 23 x 19 in. |
Length (Metric) | 58.4 cm |
Width (Metric) | 48.03 cm |
Autoclavable | Autoclavable |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction