Learn More
Fisherbrand™ Blank Writable Biohazard Identity Labels


Shop All Fisherbrand Products



Description
Strong adhesive ensures that labels remain in place.
Compliance
Meet OSHA requirements.
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
Certifications/Compliance | OSHA |
Color | Red |
Format | Label |
Material | Paper |
Product Type | Biohazard Identity Label |
For Use With (Application) | Biohazard |
Quantity | 640/Pk. |
Size (English) | 0.5 x 1.5 in. |
Size (Metric) | 1.27 x 3.81 cm |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.