Learn More
Fisherbrand™ Cartridge Kits for Milli-Q™ Pure Water Polishing Systems
Catalog No. p-169204 Shop All Fisherbrand Products
Click to view available options
Includes:
One Carbon, two ion exchange cartridges, one hollow Fiber 0.2μm and two extra final filters for critical applications
One Carbon, two ion exchange, one organic extraction, one hollow Fiber 0.2μm and two extra final filters for critical applications
High-quality filters fit Millipore™ Milli-Q three- or four-bowl systems—without expensive hardware or conversion kits
  • Deliver bacteria-free Type I reagent-grade water for a variety of applications
  • Cartridge housing: polypropylene
  • Ultrasonically welded
  • Ink-, solvent- and dye-free
  • Choose from multicartridge kits with complete single or double change
  • Double-change kits include two extra final filters for critical applications
TRUSTED_SUSTAINABILITY
Specifications
Material Polypropylene
For Use With (Equipment) Milli-Q™ pure water polishing system
Includes One Carbon, two ion exchange, one organic extraction, one hollow Fiber 0.2μm and two extra final filters for critical applications
Type Cartridge Kit
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.