Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisherbrand™ Caution: Contains Formaldehyde Labels
GREENER_CHOICESDP

Catalog No. 18999928 Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
320/Pk.
640/Pk.
Size (English):
0.5 x 1.5 in.
0.87 x 3 in.
Size (Metric):
1.27 x 3.81 cm
2.2 x 7.62 cm
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Size (English) Size (Metric)
18-999-928 320/Pk. 0.87 x 3 in. 2.2 x 7.62 cm
18-999-929 640/Pk. 0.5 x 1.5 in. 1.27 x 3.81 cm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 18-999-928
Only null left
Add to Cart
Add to Cart

Bright red fluorescent labels display the universal biohazard symbol for easy identification.

Strong adhesive ensures that labels remain in place.

Compliance

Meet OSHA requirements.

Specifications

Color White, Black, Red, Fluorescent
Format Label
Material Paper
Product Type Biohazard Warning Tape
Display Text Contains Formaldehylde
For Use With (Application) Biohazard
Quantity 320/Pk.
Size (English) 0.87 x 3 in.
Size (Metric) 2.2 x 7.62 cm
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.