Learn More
Fisherbrand™ Chemical Splash Goggles
SCI_ED
Catalog No. s01817 Shop All Fisherbrand Products
Click to view available options
:
PC lens w/antifog, antiscratch coating
This item is not returnable. View return policy
Catalog No. S01817
Item Discontinued This item has been discontinued by the Fisher Scientific channel. Please to view product availability in your area.
View Alternate Product
This item is not returnable. View return policy
Lens coated to minimize scratching and fogging.
  • Built for comfortable protection with a soft, flexible PVC body that conforms to facial contours
  • Large frames fit over most prescription eyewear
  • Coated, clear polycarbonate lens resists impact, scratching and fogging
  • Headstrap easily adjusts for a custom fit
  • Resist a wide range of chemicals and feature indirect ventilation to direct airflow over the lens
Compliance
Meet ANSI™ Z87.1-2003
Specifications
Lens Coating Anti-fog, Scratch-resistant
Certifications/Compliance ANSI Z87.1-2003
Lens Tint Clear
Product Type Chemical Splash Protection Goggles
Ventilation Details Four indirect vents
Material Polycarbonate
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.