Learn More
Fisherbrand™ Contempo Safety Glasses
Catalog No. 191302093 Shop All Fisherbrand Products
Click to view available options
Frame Color:
Black
Blue
Lens Tint:
Clear
Tinted
This item is not returnable. View return policy
Catalog No. 19-130-2093
This item is not returnable. View return policy
Offer single-lens styling with a dual-lens look
  • Polycarbonate, high-visibility lens absorbs >99.9% of harmful UV light and includes a permanent antifog coating
  • Wraparound frame with sideshields and browguard
  • Adjustable temple lengths
  • Lightweight
Compliance
Meet ANSI™ Z87.1-2010 standards.
Specifications
Lens Coating Anti-fog
Certifications/Compliance Meets ANSI Z87.1-2003 standards.
Description Tinted
Lens Tint Tinted
Frame Color Black
Temple Style Adjustable length
Lens Material Polycarbonate
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.