Learn More
Hellma Cylindrical Cells
Fit standard cell holders
Specifications
Material Suprasil
View More Specs
Products 1
Catalog Number Path Length Price Quantity & Availability  
Catalog Number Path Length Price Quantity & Availability  
14-385-862
View Documents
2 mm N/A N/A N/A
Description
Description
Cylindrical cells have a filler neck and tapered PTFE stopper (two necks and stoppers on 50 and 100mm lightpath cells).
  • Uniform diameter of 22mm
  • Available with Suprasil™ quartz or near-UV glass windows
Specifications
Specifications
Suprasil
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.