Fisherbrand™ Disposable Inoculating Loops and Needles, Length: 8 in. (20cm)

Designed for quantitative procedures such as sampling, serial dilutions, as well as for bacterial inoculation. Fisherbrand™ Disposable Inoculating Loops and Needles, Length: 8 in. (20cm) are precision molded from a specially formulated, biologically inert polymer.

 View more versions of this product

Catalog No. 22-363-605

  Check Availability
Add to Cart

    Specifications

    No. per Case 250
    Packaging Type Paper Peel Pouch, Individually wrapped
    Type Rigid Inoculating Loop and Needle
    No. per Pack 1
    Length (English) 8 in.
    Length (Metric) 20cm
    Diameter (English) Inside 0.14 in.
    Diameter (Metric) Inside 0.38cm
    Disposable Yes
    Includes Certificate of dimensional accuracy
    Volume (Metric) 10μL
    Material Biologically Inert Polymer
    Packaging Quantity 250/Cs.
    Sterile Yes
    Color Yellow

    Ideal for anaerobic microbiology applications where deep agar inoculation is required. Use in situations where flaming is not practical, such as under hoods and in anaerobic chambers. Flexible enough to facilitate streaking, yet rigid enough to penetrate the media for subsurface inoculation.

     
    • Combi Loop™ has a 10µL loop on one end and a 1µL loop on the other
    • Convenient than metal loops; use a loop directly from the package, then throw it away
    • Gamma irradiated for sterility in accordance with U.S. Pharmacopeia Volume XX
    • No flaming required, eliminating the hazards of open flames, spattering, cross-contamination, and the risk of infection from pathogens distributed by aerosol formation
    • Color coded for easy identification

  • Description & Specifications
  • Promotions

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit